Learn More
Silicon(IV) nitride, alpha-phase, ca. 90%, Thermo Scientific Chemicals
65.43 € - 517.58 €
Chemical Identifiers
CAS | 12033-89-5 |
---|---|
Molecular Formula | N4Si3 |
Molecular Weight (g/mol) | 140.28 |
MDL Number | MFCD00011230 |
InChI Key | HQVNEWCFYHHQES-UHFFFAOYSA-N |
Synonym | silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous |
PubChem CID | 3084099 |
IUPAC Name | Silicon nitride |
SMILES | N12[Si]34N5[Si]11N3[Si]25N41 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11310228
|
Thermo Scientific Alfa Aesar
042949.18 |
50 g |
65.43 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11300228
|
Thermo Scientific Alfa Aesar
042949.30 |
250 g |
202.01 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320228
|
Thermo Scientific Alfa Aesar
042949.A1 |
1 kg |
517.58 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silicon(IV) nitride is used in the automobile industry as a material for engine parts such as diesel engines and glowplugs. It is used for rocker arm pads for lower wear in spark-ignition engines. Ceramics made from this show good shock resistance. Similarly bearings made from this find application in the main engines of NASA’s space shuttle. It acts as an insulator and in the manufacture of integrated circuits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12033-89-5 | |
140.28 | |
HQVNEWCFYHHQES-UHFFFAOYSA-N | |
3084099 | |
N12[Si]34N5[Si]11N3[Si]25N41 |
N4Si3 | |
MFCD00011230 | |
silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous | |
Silicon nitride |
Specifications
1900°C (sublimation) | |
Gray | |
N4Si3 | |
14,8497 | |
Insoluble in water. | |
N12[Si]34N5[Si]11N3[Si]25N41 | |
140.28 | |
140.28 | |
Si3N4 85% min (alpha form) | |
3.2 g/mL |
12033-89-5 | |
Powder | |
MFCD00011230 | |
silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous | |
HQVNEWCFYHHQES-UHFFFAOYSA-N | |
Silicon nitride | |
3084099 | |
95% | |
Odorless | |
Silicon (IV) nitride, alpha-phase |
Safety and Handling
missing translation for 'einecsNumber' : 234-796-8
missing translation for 'rtecsNumber' : VW0650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only