missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) nitride, alpha-phase, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11367008
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Silicon(IV) nitride is used in automobile industry as a material for engine parts such as diesel engines and glowplugs. It is used for rocker arm pads for lower wear in spark-ignition engines. Ceramics made from Silicon(IV) nitride show good shock resistance. Similarly, bearings made from this find application in the main engines of NASA’s space shuttle. It acts as an insulator and is used in the manufacturing of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12033-89-5
Molecular Formula N4Si3
Molecular Weight (g/mol) 140.28
MDL Number MFCD00011230
InChI Key HQVNEWCFYHHQES-UHFFFAOYSA-N
Synonym silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
PubChem CID 3084099
IUPAC Name Silicon nitride
SMILES N12[Si]34N5[Si]11N3[Si]25N41

Specifications

Melting Point 1900°C (sublimation)
Color Gray
Physical Form Powder
Quantity 500 g
Merck Index 14,8497
Solubility Information Insoluble water.
Formula Weight 140.28
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Si3N4 90% min. (alpha form)
Odor Odorless
Density 3.2 g/mL
Assay (metals basis)
Chemical Name or Material Silicon(IV) nitride, alpha-phase
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.