missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon(IV) nitride, MgO binder, sputtering target, 76.2mm (3.0in.) dia x 6.35mm (0.250in.) thick, 99.9% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
1 Ea.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Quantity | 1 Ea. |
MDL Number | MFCD00011230 |
Formula Weight | 140.28 |
Percent Purity | 99.9% |
Assay | (metals basis) |
Chemical Name or Material | Silicon(IV) nitride, MgO binder, Sputtering Target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'