Learn More
Silicon(IV) oxide, 15% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
High-Temperature Binders, Anti-Slip Coatings For Paper, Petrochemical Catalyst Support and Binder
61.90 € - 130.00 €
Chemical Identifiers
CAS | 7631-86-9 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334187
|
Thermo Scientific Alfa Aesar
036669.A1 |
1 kg |
61.90 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11324187
|
Thermo Scientific Alfa Aesar
036669.A7 |
5 kg |
130.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
High-temperature binders, anti-slip coatings for paper, petrochemical catalyst support and binder, polishing agent, abrasion resistant coatingsSilicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of food and pharmaceutical products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
S.A. 650 m2/gChemical Identifiers
7631-86-9 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
O=[Si]=O |
O2Si | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 |
Specifications
7631-86-9 | |
O2Si | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.09 | |
Silicon(IV) oxide, 15% in H2O |
Particles in Liquid | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
Immiscible with water. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
15% |
Safety and Handling
H373: May cause damage to organs through prolonged or repeated exposure.
missing translation for 'einecsNumber' : 231-545-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only