Learn More
Silicon(IV) oxide, 40% in H{2}O, colloidal dispersion, Thermo Scientific Chemicals
Description
Functionalized RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene; acrylate and acrylamide monomers. Azide group can be used to conjugate to a variety of alkyne-functionalized biomolecules. Chain Transfer Agent (CTA). SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. Used as a suspending and thickening agent in pharmaceutical
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless |
Physical Form | Particles in Liquid |
Quantity | 5 kg |
Solubility Information | Insoluble in water. |
IUPAC Name | dioxosilane |
Formula Weight | 60.09 |
Odor | Odorless |
Density | 1.3 |
Chemical Name or Material | Silicon(IV) oxide, 40% in H2O |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.