missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

125.63 € - 340.70 €

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11390348
View Documents
Thermo Scientific Alfa Aesar
043736.14
25 g
125.63 €
25g
Please to purchase this item. Need a web account? Register with us today!
11300358
View Documents
Thermo Scientific Alfa Aesar
043736.22
100 g
340.70 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Silica is used primarily in the production of glass for windows, drinking glasses, beverage bottles. The majority of optical fibers for telecommunication are also made from silica. It is a primary raw material for many ceramics such as earthenware, stoneware, and porcelain. The main ingredient in sand casting for the manufacture of a large number of metallic components in engineering and other applications. Silica is a common additive in the production of foods. An estimated 95% of silicon dioxide produced is consumed in the construction industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7631-86-9
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O

Specifications

2230°C
7631-86-9
Fused Lump
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
Insoluble in water.
O=[Si]=O
60.08
CHEBI:30563
Odorless
Silicon(IV) oxide
1710°C
White
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
2.2-2.6
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-545-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only