Learn More
Silicon(IV) oxide, Amorphous fumed, Surface treated, S.A. 105 to 130m2/g, Thermo Scientific Chemicals
Description
Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 1 kg |
Solubility Information | Insoluble in water. |
IUPAC Name | dioxosilane |
Formula Weight | 60.09 |
Odor | Odorless |
Density | 1.8 |
Chemical Name or Material | Silicon(IV) oxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.