Learn More
Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7631-86-9 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410051
|
Thermo Scientific Alfa Aesar
044781.14 |
25 g |
N/A
|
N/A | |||||
11400051
|
Thermo Scientific Alfa Aesar
044781.22 |
100 g |
N/A
|
N/A | |||||
11420051
|
Thermo Scientific Alfa Aesar
044781.36 |
500 g |
N/A
|
N/A | |||||
Description
Silicon(IV) oxide is used as sodium aluminate stabilizer. It is also used as SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. It is also acts as chain transfer agent in polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7631-86-9 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
2230°C | |
7631-86-9 | |
Powder | |
O2Si | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.09 | |
2.4 |
1710°C | |
White | |
99.9% (metals basis) | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
Insoluble in water. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Odorless | |
Silicon(IV) oxide |
Safety and Handling
Wear suitable protective clothing.
P260-P314-P501c
H373
missing translation for 'einecsNumber' : 231-545-4
missing translation for 'rtecsNumber' : VV7565000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Silicon(IV) oxide, amorphous, nanopowder
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Silica, amorphous
RUO – Research Use Only