missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

240.20 € - 716.57 €

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357007
View Documents
Thermo Scientific Alfa Aesar
010856.14
25 g
240.20 €
25g
Please to purchase this item. Need a web account? Register with us today!
11367007
View Documents
Thermo Scientific Alfa Aesar
010856.22
100 g
716.57 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Silica is used in the manufacture of glass for windows, drinking glasses, beverage bottles, and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware, and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14808-60-7
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
O=[Si]=O
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563

Specifications

2230°C
14808-60-7
Powder
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
24261
60.09
Silicon(IV) oxide
1710°C
Brown to White
99.999% (metals basis)
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
Silica is poorly soluble in water. The solubility is higher for the amorphous form compared to the crystalline compound.
O=[Si]=O
60.08
CHEBI:30563
Odorless
Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P281-P308+P313-P501c

H350-H373

missing translation for 'einecsNumber' : 238-878-4

missing translation for 'rtecsNumber' : VV7330000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.