missing translation for 'onlineSavingsMsg'
Learn More
Silicon lump, 15cm (5.9in) & down, 99.999+% (metals basis), Thermo Scientific Chemicals Product Code.: 11393468

Silicon lump, 15cm (5.9in) & down, 99.999+% (metals basis), Thermo Scientific Chemicals

Product Code. 11393468
25 g
Click to view available options
Quantity:
25 g
100 g
1 kg
This item is not returnable. View return policy

Product Code. 11393468

Brand: Thermo Scientific Alfa Aesar 039101.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Quantity 25 g
Percent Purity ≥99.999%
Odor Odorless
Particle Size 15cm (5.9 in.) & down
Form Lump
Assay (metals basis)
Chemical Name or Material Silicon lump

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ04L021 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog Number039101.A4,039101.14,039101.22,039101.A1
Lot NumberM10L011 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number039101.14,039101.A4,039101.22,039101.A1
Lot NumberZ16K009 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog Number039101.14,039101.A4,039101.22,039101.A1
Lot NumberY19K024 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number039101.14,039101.A4,039101.22,039101.A1
Lot NumberS22I014 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number039194.M1,039194.K2,039194.K7,039101.14,039101.A4,039101.22,039101.A1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon lump, 15cm (5.9in) & down, 99.999+% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.