Learn More
Silicon powder, APS 1-5 micron, 99.9% (metals basis), Thermo Scientific Chemicals
106.53 € - 811.04 €
Chemical Identifiers
CAS | 7440-21-3 |
---|---|
Molecular Formula | Si |
Molecular Weight (g/mol) | 28.09 |
MDL Number | MFCD00085311 |
InChI Key | XUIMIQQOPSSXEZ-UHFFFAOYSA-N |
Synonym | metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 |
PubChem CID | 5461123 |
ChEBI | CHEBI:27573 |
IUPAC Name | silicon |
SMILES | [Si] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369678
|
Thermo Scientific Alfa Aesar
044185.09 |
10 g |
106.53 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379678
|
Thermo Scientific Alfa Aesar
044185.18 |
50 g |
219.09 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359678
|
Thermo Scientific Alfa Aesar
044185.30 |
250 g |
811.04 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-21-3 | |
28.09 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
5461123 | |
silicon |
Si | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
CHEBI:27573 | |
[Si] |
Specifications
7440-21-3 | |
99.9% (Metals basis) | |
MFCD00085311 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
silicon | |
5461123 | |
Odorless | |
2.33g/cm3 |
Powder | |
Si | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
[Si] | |
28.09 | |
CHEBI:27573 | |
Powder | |
Silicon, For analysis APS |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-130-8
missing translation for 'rtecsNumber' : VW0400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Silicon powder, nodular
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : Silicon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.