missing translation for 'onlineSavingsMsg'
Learn More

Silicon powder, crystalline, APS ∣<50nm, 98%, Laser synthesized from vapor phase, Thermo Scientific Chemicals

Form: S.A. 70 to 100m2/g

78.69 € - 1188.92 €

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430710
View Documents
Thermo Scientific Alfa Aesar
044384.04
2 g
78.69 €
2g
Please to purchase this item. Need a web account? Register with us today!
11498560
View Documents
Thermo Scientific Alfa Aesar
044384.09
10 g
288.44 €
10g
Please to purchase this item. Need a web account? Register with us today!
11488560
View Documents
Thermo Scientific Alfa Aesar
044384.18
50 g
1188.92 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-21-3
28.09
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
5461123
silicon
Si
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
CHEBI:27573
[Si]

Specifications

7440-21-3
98%
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
5461123
Odorless
Powder
Silicon
Crystalline Powder
Si
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
[Si]
28.09
CHEBI:27573
Air Sensitive
2.33g/cm3
Videos
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : DOT Class: 4.1, Packaging Group: III

missing translation for 'einecsNumber' : 231-130-8

missing translation for 'rtecsNumber' : VW0400000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.