missing translation for 'onlineSavingsMsg'
Learn More
Silicon rod, polycrystalline, unpolished, 1.6cm (0.6in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals Product Code.: 11387818

Silicon rod, polycrystalline, unpolished, 1.6cm (0.6in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11387818
100 mm
Click to view available options
Quantity:
10 mm
100 mm
This item is not returnable. View return policy

Product Code. 11387818

Brand: Thermo Scientific Alfa Aesar 036376.DM

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Quantity 100 mm
Percent Purity 99.9999%
Odor Odorless
Weight ≈4.68g/cm
Form Polycrystalline
Assay (metals basis)
Chemical Name or Material Silicon rod, Unpolished, 1.6cm (0.6 in.) dia.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon rod, polycrystalline, unpolished, 1.6cm (0.6in) dia, 99.9999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.