missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon Standard For ICP, 1000μg/mL in H2O/0.24 F, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
60.80 € - 129.00 €
Specifications
Chemical Name or Material | Silicon Standard |
---|---|
CAS | 16919-19-0, 7732-18-5 |
Packaging | Triple-leached LDPE bottle |
CAS Min % | 0.1, 99.9 |
CAS Max % | 0.1, 99.9 |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11963703
|
SPEX Certiprep
PLSI9-2Y |
125 mL | Triple-leached LDPE bottle |
60.80 €
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10325792
|
SPEX Certiprep
PLSI9-2X |
500 mL | Triple-leached LDPE bottle |
129.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Si | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Silicon Standard | |
Triple-leached LDPE bottle | |
0.1, 99.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
F6H8N2Si | |
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F | |
178.153 | |
Assurance | |
H2O/0.4% F |
16919-19-0, 7732-18-5 | |
0.1, 99.9 | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
diazanium;hexafluorosilicon(2-) | |
28145 | |
Matrix Concentration Cat. No.H2O/tr. HF 1000μg/mL PLSI9-2Y, -2T, -2X 10,000 PLSI9-3Y, -3XH2O 1000μg/mL PLSI9A-2X 10,000 PLSI9A-3X |
Safety and Handling
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only