Learn More
SI Analytics™ Silicon Tube/Cable Combination
Spare part for measuring stands and viscometers
Brand:  SI Analytics™ 285414031
Product Code. 10131671
  • 348.00 € / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Connect viscometers of type: 517, 520, 530, 532, 537, 538, 547, 548 and 549 to a AVS 310, 350 and 450
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tube/cable combination for viscometers
Silicon
Connect Viscometers of Type: 517, 520, 530, 532, 537, 538, 547, 548 and 549 to a AVS 310, 350 and 450