Learn More
BINDER™ Door Gasket
Door gasket - made of FKM, silicone-free
Brand: BINDER™ 8612-0007
This item is not returnable.
View return policy
Description
- Perfectly matched, durable gasket
Specifications
Door gasket - made of FKM, silicone-free | |
BINDER Chambers VD 23, VDL 23 | |
FKM Fluorocarbon | |
Door Gasket |