missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Silicone-free Viton™ Door Sealing for Thermo Scientific™ Heratherm™ Incubators and Ovens
For use with Thermo Scientific™ Heratherm™ Incubators and Ovens
824.10 € - 1648.20 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12625326
|
Thermo Scientific™
50135870 |
Heratherm 750l Models |
1648.20 €
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
|||||
12615326
|
Thermo Scientific™
50135869 |
Heratherm 400l Models |
824.10 €
Each |
Estimated Shipment: 27-03-2025
Log in to see stock. |
|||||
Description
Specifications
Door Seal |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title