Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
Brand:  Deutsch Neumann™ 1010514
 View more versions of this product
Product Code. 10281631
  • 0.50 € / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12 871
  • Durometer 50 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
14 mm
18 mm
Silicone
Silicone Stopper