Learn More
DWK Life Sciences Silicone Sealing Ring, VMQ
The silicone seal is used for sealing and fixing threaded pipes with threaded caps.
45.65 € - 192.00 €
Specifications
Material | Silicone (VMQ) |
---|---|
For Use With (Application) | Sealing; Connecting glass tubes; Suitable for PBT screw cap with aperture |
Quantity | 10/Pk. |
Product Code | Brand | Diameter (Metric) Inside | Joint Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Joint Size | Price | Quantity & Availability | ||||
11777118
|
DWK Life Sciences
292382602 |
25.0 to 27.0 mm | GL 45 |
192.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11831010
|
DWK Life Sciences
292371202 |
11.0 to 13.0 mm | GL 25 |
73.60 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11786828
|
DWK Life Sciences
292350804 |
7.5 to 9.0 mm | GL 18 |
45.65 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11747118
|
DWK Life Sciences
292340606 |
5.5 to 6.5 mm | GL 14 |
45.90 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11338803
|
DWK Life Sciences
292370806 |
7.5 to 9.0 mm | GL 25 |
73.60 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11757198
|
DWK Life Sciences
292371005 |
9.0 to 11.0 mm | GL 25 |
73.60 €
Pack of 10 |
Estimated Shipment: 09-01-2025
Log in to see stock. |
||||
11777198
|
DWK Life Sciences
292351003 |
9.0 to 11.0 mm | GL 18 |
56.95 €
Pack of 10 |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
12851054
|
DWK Life Sciences
292361407 |
13.0 to 15.0 mm | GL 32 |
112.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
13157444
|
DWK Life Sciences
292361604 |
15.0 to 17.0 mm | GL 32 |
105.00 €
Pack of 10 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
13118043
|
DWK Life Sciences
292361801 |
17.0 to 19.0 mm | GL 32 |
106.00 €
Pack of 10 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11331485
|
DWK Life Sciences
292383204 |
31.0 to 33.0 mm | GL 45 |
181.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11820720
|
DWK Life Sciences
292350607 |
5.5 to 6.5 mm | GL 18 |
52.40 €
Pack of 10 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
||||
Description
The silicone sealing ring matches the screw connection piece and has good sealing properties. Due to the PTFE layer on one side of the sealing ring, there is a certain inertness of the material. The included hole allows the insertion of glass rods, olives, hoses, etc.
- With bonded PTFE face
Specifications
Silicone (VMQ) | |
Sealing; Connecting glass tubes; Suitable for PBT screw cap with aperture | |
10/Pk. |