missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Borosilicate Glass Desiccator Lid

Manufactured from borosilicate glass. BRAND™ Borosilicate Glass Dessicator Lid helps to protect hygroscopic chemicals and preserves objects that react with humidity. Fits BRAND™ Novus Desiccator 65243

Brand:  BRAND™ 65731

117.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10622632

  • 149.00 € / Each
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from borosilicate glass.
  • Protects hygroscopic chemicals and preserves objects that react with humidity.
  • Fits BRAND™ Novus Desiccator 65243
TRUSTED_SUSTAINABILITY
Specifications

Specifications

215 mm
Desiccator Lid
Borosilicate Glass
84 mm
1 Each
Lid with socket
Novus Desiccators
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Borosilicate Glass Desiccator Lid > Outer Diameter: 215mm; Height: 84mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.