missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Solid Color-Coded Silicone Stoppers

Use these silicone stoppers with a wide range of tubes and glassware. Solid Color-Coded Silicone Stoppers are available in 14 color-coded sizes and feature flexible construction for an easy fit.

Brand:  Cole-Parmer™ 06298-14

 View more versions of this product

Product Code. 11857432

  • 116.00 € / Pack of 10
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use these silicone stoppers with a wide range of tubes and glassware. Solid Color-Coded Silicone Stoppers are available in 14 color-coded sizes and feature flexible construction for an easy fit.

  • Silicone bubbles make these stoppers so lightweight they float if dropped in water
  • Softness and flexibility allow stoppers to fit a range of holes
  • Handle temperatures from –76 to 392°F (–60 to 200°C)
  • Adapt to your needs—bore holes for thermometer or tubing addition
  • Easily make openings for glassware fit your exact specifications with the optional cork borer set Autoclavable
Specifications

Specifications

10/Pk.
–60 to 200°C
33 mm
Silicone
8
Green
40 mm
Solid Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.