missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Silicone Tubing

Silicone tubing for Peristaltic Pump P-1. Cytiva Silicone Tubing with i.d. 3.1 mm.

Brand:  Cytiva 19-4690-01

Product Code. 11360002

  • 86.59 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Tubing, 140 mm, i.d. 3.1 mm
  • Silicone
  • Packsize 2 pieces
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tubing
1 Each
140 mm
3.1 mm
For P-1 Peristaltic Pump
Silicone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers