missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Silicone Tubing
Durable silicone tubing well suited for a variety of pump and transfer applications.
Brand: Brand™ 143357
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Translucent
- Autoclavable (121°C)
- Note: limited chemical resistance when used with concentrated acids
- Minimum order of 25 meters
Specifica
Tubing | |
4 mm | |
7 mm | |
Translucent |
Autoclavable | |
Silicone | |
1.5 mm |