Learn More
Thermo Scientific™ SilTite™ Ferrule Kits
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits.
Brand:  Thermo Scientific™ 290MA217
 View more versions of this product
Product Code. 10086154
  • 193.12 € / Pack of 10
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ferrule Kit
SilTite Kit for Thermo Scientific TRACE 1300/1310 GC SSL Injectors
Two SilTite Nuts and Ten Ferrules
290 MA217
SilTite Metal
0.53 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.