missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ SilTite™ Ferrules

Ensures easy, leak-free installation for capillary columns without the use of any tools.

76.40 € - 118.00 €


Products 5
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
15218923
View Documents Promotion Details
Trajan Scientific and Medical™
073228
For Use with Shimadzu Detector/Injectors, Standard MS Interface 0.32 mm I.D. Columns
118.00 €
Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
15288923
View Documents Promotion Details
Trajan Scientific and Medical™
073332
For Use with Thermo GC/MS Interface Connections 0.45 to 0.53 mm I.D. Columns
76.40 €
Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
15228923
View Documents Promotion Details
Trajan Scientific and Medical™
073229
For Use with Shimadzu Detector/Injectors, Standard MS Interface 0.45 to 0.53 mm I.D. Columns
118.00 €
Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
15278923
View Documents Promotion Details
Trajan Scientific and Medical™
073331
For Use with Thermo GC/MS Interface Connections 0.32 mm I.D. Columns
76.40 €
Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
15268923
View Documents Promotion Details
Trajan Scientific and Medical™
073330
For Use with Thermo GC/MS Interface Connections 0.10 to 0.25 mm I.D. Columns
76.40 €
Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
Description

Description

  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Videos
SDS
Documents

Documents

Product Certifications