Learn More
Uvex Silv-Air Classic 3100 FFP1 Mask
The uvex silv-Air c product range consists of both classic folding and cup-style respirators that fit a wide variety of facial shapes.
Brand:  Uvex 8763100
32.51 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 17905446
  • 41.24 € / Pack of 30
Explore more special offers
Description
Description
  • With headband loops and integrated sealing lips in all protection classes, it is really easy to fit the masks correctly.
  • The masks can also be worn comfortably for a longer period of time without becoming bothersome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P - Oil-Proof
Flat-fold
KN95
General Purpose Particulate Respirator
No
Sterile
White
Reusable
Polypropylene, Polyester
Medium
Yes