missing translation for 'onlineSavingsMsg'
Learn More

Silver acetate, anhydrous, 99%, Thermo Scientific Chemicals

Product Code. 11311778
Click to view available options
Quantity:
25 g
100 g

It is a reagent in the laboratory as a source of silver ions lacking an oxidizing anion. It is a reagent for direct ortho-arylation, and for conversion of organohalogen compounds into alcohols. Woodward cis-hydroxylation reaction employs silver acetate and iodine for selective conversin of alkenes into cis-diols. Silver acetate is the more preferred reagent for facile carbonylation of primary and secondary amines. It is also employed in the preparation of highly reflective, conductive silvered polymer films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-63-3
Molecular Formula C2H3AgO2
Molecular Weight (g/mol) 166.912
MDL Number MFCD00012446
InChI Key CQLFBEKRDQMJLZ-UHFFFAOYSA-M
Synonym silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
PubChem CID 11246
IUPAC Name silver;acetate
SMILES CC(=O)[O-].[Ag+]

Specifications

Melting Point (decomposition)
Color White to Gray
Physical Form Crystalline
Quantity 25 g
Linear Formula AgOOCCH3
UN Number UN3077
Beilstein EM>10,
Merck Index 14,8505
Solubility Information Soluble in dilute HNO3; Solubility in water increases with temperature.
Formula Weight 166.92
Percent Purity 99%
Odor Odorless
Sensitivity Light sensitive
Density 3.26 g/mL
Chemical Name or Material Silver acetate, Anhydrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver acetate, anhydrous
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Silver acetate, anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.