missing translation for 'onlineSavingsMsg'
Learn More

Silver benzoate, 99%, Thermo Scientific Chemicals

Product Code. 10421641
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-31-0
Molecular Formula C7H5AgO2
Molecular Weight (g/mol) 228.98
MDL Number MFCD00013030
InChI Key CLDWGXZGFUNWKB-UHFFFAOYSA-M
Synonym silver benzoate hydrate
PubChem CID 87384783
IUPAC Name benzoic acid;silver;hydrate
SMILES [Ag+].[O-]C(=O)C1=CC=CC=C1

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CO2Ag
Quantity 50 g
Merck Index 15, 1093
Solubility Information Solubility in water: 0.26%. Other solubilities: slightly soluble in alcohol
Formula Weight 228.99
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Silver benzoate, 99%
Show More Show Less
Product Identifier
  • Silver benzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'