missing translation for 'onlineSavingsMsg'
Learn More

Silver bromide, 99.5%, Thermo Scientific Chemicals

Product Code. 11301259
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Silver bromide is used as the light-sensitive component on ordinary photographic film and plates and also used in I.R. applications and ionic semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-23-1
Molecular Formula AgBr
Molecular Weight (g/mol) 187.772
MDL Number MFCD00003398
InChI Key ADZWSOLPGZMUMY-UHFFFAOYSA-M
Synonym silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion
PubChem CID 66199
IUPAC Name bromosilver
SMILES Br[Ag]

Specifications

Melting Point 432°C
Color Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9%
Solubility Information Insoluble in water,alcohol and acids. Partially soluble in ammonia.
Formula Weight 187.78
Odor Odorless
Sensitivity Light sensitive
Appearance Pale yellow powder/flake/crystalline/beads
Density 6.47 g/cm3 at 20°C
Chemical Name or Material Silver bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'