missing translation for 'onlineSavingsMsg'
Learn More

Silver carbonate, 99%, pure

Product Code. 10684862 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10684862

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 534-16-7 | CAg2O3 | 275.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-16-7
Molecular Formula CAg2O3
Molecular Weight (g/mol) 275.75
MDL Number MFCD00003403
InChI Key WIKQEUJFZPCFNJ-UHFFFAOYSA-N
Synonym carbonic acid; silver, carbonic acid disilver
PubChem CID 25203940
IUPAC Name carbonic acid;silver
SMILES C(=O)(O)O.[Ag].[Ag]

Specifications

Melting Point 210.0°C
Color Green to Green-Yellow
Physical Form Granular Powder
Quantity 25 g
Assay Percent Range 99%
Linear Formula Ag2CO3
Fieser 01,1005; 02,363; 04,425; 06,511; 08,411; 12,432
Merck Index 15, 8646
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonium hydroxide and nitric acid, insoluble in alcohol
Formula Weight 275.75
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Silver carbonate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver carbonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.