missing translation for 'onlineSavingsMsg'
Learn More
Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11325869

Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11325869
50 x 50 mm
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
200 x 200 mm
This item is not returnable. View return policy

Product Code. 11325869

Brand: Thermo Scientific Alfa Aesar 011440.FI

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 50 x 50 mm
Percent Purity 99.9%
Odor Characteristic
Weight ≈3.3g/50x50mm
Form Foil
Assay (metals basis)
Chemical Name or Material Silver foil, 0.127mm (0.005 in.) thick, Annealed, Temper: soft

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.