missing translation for 'onlineSavingsMsg'
Learn More
Silver foil, 0.25mm (0.01in) thick, hard, Premion, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11385909

Silver foil, 0.25mm (0.01in) thick, hard, Premion, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11385909
50 x 50 mm
missing translation for 'orderingAttributeHoverText'
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
This item is not returnable. View return policy

Product Code. 11385909

Brand: Thermo Scientific Alfa Aesar 012125.FI

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 6.6g/50 x 50mm; Temper: as rolled

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored foil
Density 10.5g/cm3
Assay Percent Range 99.998% (Metals basis)
Quantity 50 x 50 mm
Odor Characteristic
Form Foil
Chemical Name or Material Silver, Hard

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY18I035 Certificate TypeCertificate of Analysis Date25/01/2023 Catalog Number012125.GH,012125.FI,012125.HP
Lot NumberW28D060 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012125.GH,012125.FI,012125.HP
Lot NumberI27Z025 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012125.GH,012125.FI,012125.HP
Lot NumberI16U009 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number012125.GH,012125.FI,012125.HP
Lot NumberN22C039 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number012125.GH,012125.FI,012125.HP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver foil, 0.25mm (0.01in) thick, hard, Premion, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.