missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver methanesulfonate, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Silver methanesulfonate is used as a catalyst for heterocyclization reactions. Further, it is used in the synthesis of unsaturated ketones and esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 10 g |
Solubility Information | Soluble in water,alcohol,ether and benzene. Slightly soluble in toluene,o-chlorotoluene and ethyl disulfide. Insoluble in hexane and methylcyclopentane. |
Formula Weight | 202.97 |
Odor | Odorless |
Sensitivity | Light sensitive |
Appearance | White powder |
Chemical Name or Material | Silver methanesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'