missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate, 0.1 N standard solution, Thermo Scientific Chemicals

104.52 € - 333.66 €

Specifications

Chemical Name or Material Silver nitrate
Name Note 0.1 N Standard Solution
CAS 7761-88-8, 7732-18-5
Color Colorless
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10487700
View Documents
Thermo Scientific Acros
124250010
1 L HDPE Bottle
104.52 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10512734
View Documents
Thermo Scientific Acros
124250050
5 L HDPE Bottle
333.66 €
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
Specifications

Specifications

Silver nitrate
7761-88-8, 7732-18-5
Liquid
AgNO3
01,1008; 02,366; 03,252; 04,429; 05,582; 07,321; 09,411; 10,350; 11,268; 12,257; 14,350; 15,39
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
169.87
0.0998 to 0.1002N (20°C)
1.015
0.1 N Standard Solution
Colorless
AgNO3
MFCD00003414
15, 8657
Solubility in water: miscible.
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
Pure
HDPE Bottle
1.0150g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 231-853-9

Product Identifier
  • Silver nitrate, 0.1N standard solution

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only