Learn More
Silver nitrate, 99.85%, for analysis, Thermo Scientific Chemicals
50.65 € - 5052.14 €
Chemical Identifiers
CAS | 7761-88-8 |
---|---|
Molecular Formula | AgNO3 |
Molecular Weight (g/mol) | 169.87 |
MDL Number | MFCD00003414 |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
PubChem CID | 24470 |
ChEBI | CHEBI:32130 |
SMILES | [Ag+].[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10503951
|
Thermo Scientific Acros
197680050 |
5 g | Glass bottle |
50.65 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10449900
|
Thermo Scientific Acros
197680250 |
25 g | Glass bottle |
127.64 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10164260
|
Thermo Scientific Acros
197681000 |
100 g | Glass bottle |
429.14 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10684892
|
Thermo Scientific Acros
197685000 |
500 g | Glass bottle |
1445.19 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974341
|
Thermo Scientific Acros
197680025 |
2.5 kg | Glass Bottle |
5052.14 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisChemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
[Ag+].[O-][N+]([O-])=O |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 |
Specifications
7761-88-8 | |
Crystalline Powder and/or Crystals | |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
≥99.80% | |
0.1% (1g, 70°C, 2 hrs) max. | |
Silver nitrate |
White | |
99.85% | |
AgNO3 | |
15, 8657 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver;nitrate | |
24470 | |
169.87 | |
Analytical | |
>444°C |
Safety and Handling
GHS P Statement
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only