missing translation for 'onlineSavingsMsg'
Learn More
Silver nitrate, 99+%, Thermo Scientific Chemicals Product Code.: 11450990

Silver nitrate, 99+%, Thermo Scientific Chemicals

Product Code. 11450990
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11450990

Brand: Thermo Scientific Alfa Aesar A13854.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silver nitrate is employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology. It finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions. Silver(I) nitrate can be used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis. It is widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Employed in Silver plating, photography, manufacturing of other silver compounds, mirrors, coloring porcelain, and staining in histology.
  • Finds use as a stain in scanning electron microscopy, and a key ingredient in several silver-based antiseptic and antibacterial pharmaceutical compositions.
  • Used to separate mixtures of alkenes by selective absorption, and for deprotection and oxidation reactions in organic synthesis.
  • Widely used as an analytical reagent owing to its immediate reaction with halide ions forming insoluble precipitate of silver(I) halides
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O

Specifications

Boiling Point 444°C
Melting Point 212°C
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99+%
UN Number UN1493
Merck Index 14,8518
Solubility Information Freely soluble aq. soln., alcohol and aqueous ammonia. Slightly soluble in ether.
IUPAC Name silver(1+) nitrate
Formula Weight 169.87
Packaging Glass jar
Sensitivity Light Sensitive
Appearance White
Decomposition Information >444°C
Density 4.352
Chemical Name or Material Silver nitrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62480080 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog NumberA13854.22,A13854.14,A13854.36,A13854.06
Lot Number10252994 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog NumberA13854.22,A13854.14,A13854.36,A13854.06
Lot Number10219658 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog NumberA13854.22,A13854.14,A13854.36,A13854.06
Lot Number10237365 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13854.22,A13854.14,A13854.36,A13854.06
Lot Number10T021 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13854.22,A13854.14,A13854.36,A13854.06
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Silver nitrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : silver nitrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver nitrate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.