missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver nitrate concentrate, for 1L standard solution, 0.01 M AgNO3, Honeywell™
Click to view available options
Quantity:
1 Pc.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS | 7761-88-8 |
Quantity | 1 Pc. |
Molecular Formula | AgNO3 |
MDL Number | MFCD00003414 |
UN Number | UN3082 |
Synonym | silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
SMILES | [Ag+].[O-][N+]([O-])=O |
IUPAC Name | silver(1+) nitrate |
Molecular Weight (g/mol) | 169.87 |
Show More |
Safety and Handling
Product Identifier
- Silver nitrate concentrate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : silver nitrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction