missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate Solution, 0.01M, Honeywell™

Product Code. 15634160
Click to view available options
Quantity:
1 L

Volumetric, 0.01 M AgNO3 (0.01N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7761-88-8
Quantity 1 L
Molecular Formula AgNO3
MDL Number MFCD00003414
UN Number UN3082
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87g/mol
Concentration or Composition (by Analyte or Components) 0.01M
Chemical Name or Material Silver nitrate solution
Show More Show Less
compliance-icons
Product Identifier
  • Silver nitrate solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Silver nitrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'