missing translation for 'onlineSavingsMsg'
Learn More

Silver nitrate solution, Volumetric, Reag. Ph. Eur., 0.1 M AgNO3 (0.1N), Honeywell Fluka™

Volumetric, Reag. Ph. Eur., 0.1 M AgNO3 (0.1N)

82.84 € - 706.69 €

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
SMILES [Ag+].[O-][N+]([O-])=O
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15615020
View Documents
Honeywell-Fluka
35375-500ML
500 mL
82.84 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15695010
View Documents
Honeywell-Fluka
35375-1L
1 L
143.42 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15605020
View Documents
Honeywell-Fluka
35375-2.5L
2.5 L
248.34 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15635020
View Documents
Honeywell-Fluka
35375-5L-VP
5 L
411.21 €
5 L
Please to purchase this item. Need a web account? Register with us today!
15645020
View Documents
Honeywell-Fluka
35375-6X1L
6 x 1 L
706.69 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15685010
View Documents
Honeywell-Fluka
35375-10L-VP
10 L
576.29 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
[Ag+].[O-][N+]([O-])=O
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130

Specifications

7761-88-8
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
0.1M
AgNO3
UN3082
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
169.87g/mol
Silver nitrate solution
Videos
Safety and Handling

Safety and Handling

P273-P280-P305 + P351 + P338-P337 + P313-P391-P501

Product Identifier
  • Silver nitrate solution

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P234-Keep only in original container.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Silver nitrate solution

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.