Learn More
Silver standard solution, for AAS, 1mg/mL Ag in 0.5N HNO3, Thermo Scientific Chemicals
20.00 € - 61.51 €
Specifications
Chemical Name or Material | Silver standard solution |
---|---|
Concentration | 997 to 1003 μg/mL of Ag (1+) in 0.5N HNO3 (20°C) |
Name Note | 1 mg/ml Ag in 0.5N HNO3 |
CAS | 7761-88-8, 7697-37-2, 7732-18-5 |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10174400
|
Thermo Scientific Acros
196301000 |
100 mL | Plastic bottle |
20.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10226540
|
Thermo Scientific Acros
196305000 |
500 mL | Plastic bottle |
61.51 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopySpecifications
Silver standard solution | |
1 mg/ml Ag in 0.5N HNO3 | |
Plastic bottle | |
AgNO3 | |
15, 8657 | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
AAS | |
-3.0°C | |
1.02 | |
Liquid |
997 to 1003 μg/mL of Ag (1+) in 0.5N HNO3 (20°C) | |
7761-88-8, 7697-37-2, 7732-18-5 | |
Colorless | |
MFCD00003414 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
107.87 | |
1.0200g/mL | |
101.0°C | |
Soluble in water |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
May be corrosive to metals.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Wash face, hands and any exposed skin thoroughly after handling.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Silver atomic absorption standard solution
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Nitric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.