missing translation for 'onlineSavingsMsg'
Learn More

Silver trifluoroacetate, 98%, Thermo Scientific Chemicals

Product Code. 11391469
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Silver trifluoroacetate is used as a Lewis acid catalyst. It is used as an important raw material for the preparation of triethylsilyl trifloroacetate and triphenylmethyl trifluoroacetate. Further, it is used as a catalyst in the preparation of 4-iodoveratrole from veratrole by reacting with iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2966-50-9
Molecular Formula C2AgF3O2
Molecular Weight (g/mol) 220.883
MDL Number MFCD00013199
InChI Key KZJPVUDYAMEDRM-UHFFFAOYSA-M
Synonym silver trifluoroacetate, silver 1+ trifluoroacetate, silver i 2,2,2-trifluoroacetate, silver mono trifluoroacetate, trifluoroacetic acid silver salt, silver 1+ ion trifluoroacetate, acetic acid, trifluoro-, silver 1+ salt, silver i trifluoroacetate, trifluoroacetic acid, silver salt, trifluoroacetic acid, silver 1+ salt
PubChem CID 76299
IUPAC Name silver;2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].[Ag+]

Specifications

Melting Point 257°C to 260°C (decomposition)
Physical Form Powder
Quantity 5 g
Assay Percent Range 98%
Solubility Information Soluble in water,ether and benzene.
Formula Weight 220.88
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Silver trifluoroacetate
compliance-icons compliance-icons
Product Identifier
  • Silver trifluoroacetate
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Silver trifluoroacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.