missing translation for 'onlineSavingsMsg'
Learn More
Silver tubing, 6.35mm (0.25in) OD, 4.35mm (0.171in) ID, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11307847

Silver tubing, 6.35mm (0.25in) OD, 4.35mm (0.171in) ID, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11307847
50 cm
Click to view available options
Quantity:
10 cm
50 cm
4 x 50 cm
This item is not returnable. View return policy

Product Code. 11307847

Brand: Thermo Scientific Alfa Aesar 014259.BW

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Quantity 50 cm
Percent Purity 99.9%
Odor Characteristic
Weight ≈18g/10cm
Assay (metals basis)
Chemical Name or Material Silver tubing, OD: 6.35mm (0.25 in.) ID: 4.35mm (0.171 in.), Temper: as drawn

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH15N27 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014259.VC,014259.BS,014259.BW
Lot NumberF02N30 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014259.VC,014259.BS,014259.BW
Lot NumberL13X001 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014259.VC,014259.BS,014259.BW
Lot NumberI26R015 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014259.VC,014259.BS,014259.BW
Lot NumberC04N15 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014259.VC,014259.BS,014259.BW

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silver tubing, 6.35mm (0.25in) OD, 4.35mm (0.171in) ID, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.