missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 0.127mm (0.005in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals

146.73 € - 494.46 €

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394178
View Documents
Thermo Scientific Alfa Aesar
041454.G9
25 m
146.73 €
25m
Please to purchase this item. Need a web account? Register with us today!
11384178
View Documents
Thermo Scientific Alfa Aesar
041454.H4
100 m
494.46 €
100m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]

Specifications

7440-22-4
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
99.9%
≈0.13g/m
(metals basis)
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
107.87
CHEBI:9141
Characteristic
Wire
Silver wire, 0.127mm (0.005 in.) dia., Hard, Temper: as drawn
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-131-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only