missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific Disposable Base Molds

Disposable Base Molds are inexpensive enough to be discarded after use, yet strong enough to be reused.

Specifications

Product Type Base Mold
Material PVC
Depth (Metric) 0.5 cm
Depth (English) 0.196 in.
Disposable Yes
View More Specs

Description

Description

  • Specially formulated plastic material offers excellent thermal exchange
  • Smooth interior finish and rounded corners facilitate specimen removal
Specifications

Specifications

Base Mold
0.5 cm
Yes
1000/Cs.
PVC
0.196 in.
Can be use with same styles and types of cassettes and embedding rings and provides offering excellent thermal exchange
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.