Learn More
Simport™ Scientific Sample Tubes with External Threads - Graduated
With white marking area
Specifications
Closure Material Polyethylene
Autoclavable Autoclavable
Closure Type Screw Cap
Material Polypropylene
Sterility Non-sterile
View More Specs
Description
Description
  • Made of polypropylene
  • Smooth and uniform inner surface with reduced risk of contamination
  • Can be autoclaved at upto 121°C in upright position with caps loosened
  • Height of tube is with cap
  • Available in choice of self standing or with round bottom
  • Caps sold separately
  • Round bottom tubes can be can be centrifuged up to 17,000g
Specifications
Specifications
Polyethylene
Screw Cap
Non-sterile
External thread, graduations and white writing area
Autoclavable
Polypropylene
Storage and transportation of biological materials