Learn More
Simport™ Scientific SLIMSETTE™ M510SL Biopsy Processing/Embedding Cassettes in Quickload™ Sleeves
A new generation of cassettes, prepackaged for use in automatic cassette labeling instruments
Specifications
Material Acetal Polymer
Pore Style Slots
Writing Face Angle 45°
View More Specs
Description
Description
  • Specially designed to hold biopsy specimens during the embedding process
  • Made of acetal polymer
  • Transparent sleeve allows clear view of cassettes during printing
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • Lid can be opened and closed as often as necessary, and will always relock securely
  • Large labeling area for easy identification
  • Anterior writing area at a 45° angle
Specifications
Specifications
Acetal Polymer
Slots
45°