missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Scientific SLIMSETTE™ M511 Tissue Processing/Embedding Cassettes

A new generation of cassettes featuring four square compartments

Specifications

Closure Type Integral Lid
Material Acetal Polymer
Writing Face Angle 45°
View More Specs

Description

Description

  • Made of acetal polymer
  • Design has a unique recessed cover
  • Cassettes are thinner, allowing more to be stacked in automatic labeling machines
  • Each cassette has four square compartments measuring 13mm
  • Large labeling area for easy identification
  • Anterior writing area at a 45° angle

Order Info

3 x 500/Cs.

Specifications

Specifications

Integral Lid
Acetal Polymer
45°
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.