Learn More
Köttermann Single Bench
This Single Bench fulfills a variety of tasks in the laboratory.
Brand:  Köttermann 505-00161
 View more versions of this product
Product Code. 17382219
  • 1610.00 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available for sitting and standing work height
  • Stationary
  • Specifications
    Specifications
    750 x 1500 x 750 mm
    1
    Trespa TopLab PLUS Worktop