Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Brand: Thermo Scientific™ 840-277500
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
Specifications
Test tube and vial holder accommodating 12 mm diameter up to 25 mm diameter test tubes and vials | |
Colorimetric/spectroscopic measurements in test tube cuvettes. OD600 measurements for cell growth monitoring in tubes. Monitoring cell growth in nephelo-culture flasks. Measurements in 16 mm screw-cap vials |
|
12 mm to 25 mm diameter test tubes and vials |
Test Tube and Vial Holder | |
GENESYS 30/40/50 Spectrophotometers | |
One cuvette holder built into a sample compartment tray. Snaps into place magnetically. No tools required. |
Product Suggestions
Customers who viewed this item also viewed.