Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Brand: Thermo Scientific™ 840-303100
This item is not returnable.
View return policy
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Long path cylindrical cell holder | |
Measurements of gases and select older standard methods that call specifically for cylindrical cells | |
5 mm to 100 mm cylindrical cuvette (1) |
Long Path Cell Holder | |
GENESYS 140/150/180, BioMate 160 Spectrophotometers | |
Cylindrical |